5-Cyclopropyl-1H-pyrazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

056389-500MG 056389-1G
101910-312EA 375.38 USD
101910-312 101910-314
5-Cyclopropyl-1H-pyrazole-3-carboxylic acid ≥95%
5-Cyclopropyl-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
MDL Number: MFCD04967194
CAS Number: 890591-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR