2-[(4-Chlorophenyl)sulfanyl]-5-fluorobenzaldehyde ≥95%

Supplier: Matrix Scientific

056383-500MG 056383-1G 056383-5G
101910-286EA 278.46 USD
101910-286 101910-288 101910-290
2-[(4-Chlorophenyl)sulfanyl]-5-fluorobenzaldehyde ≥95%
2-[(4-Chlorophenyl)sulfanyl]-5-fluorobenzaldehyde

Hazard: Irritant

Formula: C₁₃H₈ClFOS
MDL Number: MFCD16710275
CAS Number: 710960-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR