5-(Chloromethyl)-1-(4-fluorophenyl)-1H-1,2,4-triazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

056379-500MG 056379-1G
101910-268EA 556.92 USD
101910-268 101910-270
5-(Chloromethyl)-1-(4-fluorophenyl)-1H-1,2,4-triazole-3-carboxylic acid ≥95%
5-(Chloromethyl)-1-(4-fluorophenyl)-1H-1,2,4-triazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₇ClFN₃O₂
MDL Number: MFCD16710302
CAS Number: 1242268-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR