2-[(2-Chloroacetyl)amino]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

056374-500MG 056374-1G
101910-248EA 181.54 USD
101910-248 101910-250
2-[(2-Chloroacetyl)amino]benzenecarboxylic acid ≥95%
2-[(2-Chloroacetyl)amino]benzenecarboxylic acid

Hazard: Irritant

Formula: C₉H₈ClNO₃
MDL Number: MFCD00029924
CAS Number: 14422-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR