2-[(2-Aminoethyl)sulfanyl]-1-methyl-1H-imidazole ≥95%

Supplier: Matrix Scientific

056368-500MG 056368-1G
101910-224EA 273.85 USD
101910-224 101910-226
2-[(2-Aminoethyl)sulfanyl]-1-methyl-1H-imidazole ≥95%
2-[(2-Aminoethyl)sulfanyl]-1-methyl-1H-imidazole

Hazard: Irritant

Formula: C₆H₁₁N₃S
MDL Number: MFCD06655922
CAS Number: 142313-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR