1-(tert-Butyl)-2-methyl-(2S,4R)-4-hydroxytetrahydro-1,2(2H)pyridinedicarboxylate ≥95%

Supplier: Matrix Scientific

056366-500MG 056366-1G
101910-216EA 921.54 USD
101910-216 101910-218
1-(tert-Butyl)-2-methyl-(2S,4R)-4-hydroxytetrahydro-1,2(2H)pyridinedicarboxylate ≥95%
1-(tert-Butyl)-2-methyl-(2S,4R)-4-hydroxytetrahydro-1,2(2H)pyridinedicarboxylate

Hazard: Irritant

Formula: C₁₂H₂₁NO₅
Melting Pt: 88…90 °C
MDL Number: MFCD02259709
CAS Number: 181269-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR