tert-Butyl-3-(chloromethyl)tetrahydro-1(2H)-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

056364-500MG 056364-1G
101910-208EA 326.15 USD
101910-208 101910-210
tert-Butyl-3-(chloromethyl)tetrahydro-1(2H)-pyridinecarboxylate ≥95%
tert-Butyl-3-(chloromethyl)tetrahydro-1(2H)-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₁H₂₀ClNO₂
MDL Number: MFCD09953348
CAS Number: 876589-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR