tert-Butyl-N-(6-chloro-4-fluoro-3-pyridinyl)carbamate ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Amino-2-chloro-4-fluoropyridine, 5-BOC protected
056363-500MG
056363-1G
101910-204EA
556.92
USD
101910-204
101910-206
tert-Butyl-N-(6-chloro-4-fluoro-3-pyridinyl)carbamate ≥95%
tert-Butyl-N-(6-chloro-4-fluoro-3-pyridinyl)carbamate
Hazard: Irritant
Formula:
C₁₀H₁₂ClFN₂O₂ MW: 246.67 g/mol |
MDL Number:
MFCD07368891 CAS Number: 915307-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...