2-[4-(tert-Butoxycarbonyl)piperazino]-5-nitronicotinic acid ≥95%

Supplier: Matrix Scientific

056361-500MG 056361-1G
101910-196EA 556.92 USD
101910-196 101910-198
2-[4-(tert-Butoxycarbonyl)piperazino]-5-nitronicotinic acid ≥95%
2-[4-(tert-Butoxycarbonyl)piperazino]-5-nitronicotinic acid

Hazard: Irritant

Formula: C₁₅H₂₀N₄O₆
MDL Number: MFCD16622803
CAS Number: 1242268-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR