6-Nitro-1,3-benzodioxol-5-amine ≥97%

Supplier: Matrix Scientific

056359-500MG 056359-1G
101910-188EA 246.15 USD
101910-188 101910-190
6-Nitro-1,3-benzodioxol-5-amine ≥97%
6-Nitro-1,3-benzodioxol-5-amine

Hazard: Irritant

Formula: C₇H₆N₂O₄
MDL Number: MFCD00184722
CAS Number: 64993-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR