Methyl-2-[4-hydroxydihydro-2(3H)-isoxazolyl]-5-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

056356-500MG 056356-1G
101910-178EA 921.54 USD
101910-178 101910-180
Methyl-2-[4-hydroxydihydro-2(3H)-isoxazolyl]-5-nitrobenzenecarboxylate ≥95%
Methyl-2-[4-hydroxydihydro-2(3H)-isoxazolyl]-5-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₆
MDL Number: MFCD16622810
CAS Number: 1242267-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR