Methyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-2-cyanoacetate ≥95%

Supplier: Matrix Scientific

056354-500MG 056354-1G 056354-5G
101910-168EA 181.54 USD
101910-168 101910-170 101910-172
Methyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-2-cyanoacetate ≥95%
Methyl-2-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-2-cyanoacetate

Hazard: Irritant

Formula: C₁₀H₆ClF₃N₂O₂
MW: 278.62 g/mol
MDL Number: MFCD16622814
CAS Number: 658066-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR