1-(3-Methoxy-4-nitrophenyl)-4-piperidinylmethanesulfonate ≥95%
Supplier: Matrix Scientific
056353-500MG
056353-1G
101910-164EA
375.38
USD
101910-164
101910-166
1-(3-Methoxy-4-nitrophenyl)-4-piperidinylmethanesulfonate ≥95%
1-(3-Methoxy-4-nitrophenyl)-4-piperidinylmethanesulfonate
Hazard: Irritant
Formula:
C₁₃H₁₈N₂O₆S |
MDL Number:
MFCD16622790 CAS Number: 1242268-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...