6-[4-Hydroxydihydro-2(3H)-isoxazolyl]nicotinonitrile ≥95%

Supplier: Matrix Scientific

056349-500MG 056349-1G
101910-146EA 921.54 USD
101910-146 101910-148
6-[4-Hydroxydihydro-2(3H)-isoxazolyl]nicotinonitrile ≥95%
6-[4-Hydroxydihydro-2(3H)-isoxazolyl]nicotinonitrile

Hazard: Irritant

Formula: C₉H₉N₃O₂
MW: 191.19 g/mol
MDL Number: MFCD16622802
CAS Number: 1242267-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR