4-Fluoroisatoic anhydride ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Fluoro-1H-benzo[d][1,3]oxazine-2,4-dione

056345-500MG 056345-1G 056345-5G
101910-126EA 278.46 USD
101910-126 101910-128 101910-130
4-Fluoroisatoic anhydride ≥95%
4-Fluoroisatoic anhydride

Hazard: Irritant

Formula: C₈H₄FNO₃
MW: 181.1200 g/mol
MDL Number: MFCD03839826
CAS Number: 321-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR