Ethyl-3-oxo-3,4-dihydro-2-quinoxalinecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-Oxo-3,4-dihydro-2-quinoxalinecarboxylate

056343-500MG 056343-1G
101910-118EA 153.85 USD
101910-118 101910-120
Ethyl-3-oxo-3,4-dihydro-2-quinoxalinecarboxylate ≥97%
Ethyl-3-oxo-3,4-dihydro-2-quinoxalinecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₃
MW: 218.21 g/mol
MDL Number: MFCD00459589
CAS Number: 36818-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR