Ethyl-6-nitroimidazo[1,2-a]pyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-nitroimidazo[1,2-a]pyridine-2-carboxylate

056342-500MG 056342-1G
101910-114EA 278.46 USD
101910-114 101910-116
Ethyl-6-nitroimidazo[1,2-a]pyridine-3-carboxylate ≥95%
Ethyl 6-nitroimidazo[1,2-a]pyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₉N₃O₄
MW: 235.2000 g/mol
MDL Number: MFCD05864803
CAS Number: 38923-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR