Ethyl-5-bromo-2-chloroisonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-bromo-2-chloroisonicotinate

056340-1G 056340-5G
101910-106EA 40 USD
101910-106 101910-108
Ethyl-5-bromo-2-chloroisonicotinate ≥95%
Ethyl-5-bromo-2-chloroisonicotinate

Hazard: Irritant

Formula: C₈H₇BrClNO₂
MW: 264.51 g/mol
MDL Number: MFCD14698168
CAS Number: 1214346-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR