(E)-3-(Dimethylamino)-1-(2-naphthyl)-2-propen-1-one ≥95%

Supplier: Matrix Scientific

056336-500MG 056336-1G 056336-5G
101910-090EA 181.54 USD
101910-090 101910-092 101910-094
(E)-3-(Dimethylamino)-1-(2-naphthyl)-2-propen-1-one ≥95%
(E)-3-(Dimethylamino)-1-(2-naphthyl)-2-propen-1-one

Hazard: Irritant

Formula: C₁₅H₁₅NO
MW: 225.29 g/mol
MDL Number: MFCD00179142
CAS Number: 138716-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR