N-(3,4-Dimethoxyphenyl)acetamide ≥95%

Supplier: Matrix Scientific

056335-500MG 056335-1G 056335-5G
101910-084EA 181.54 USD
101910-084 101910-086 101910-088
N-(3,4-Dimethoxyphenyl)acetamide ≥95%
N-(3,4-Dimethoxyphenyl)acetamide

Hazard: Irritant

Formula: C₁₀H₁₃NO₃
MDL Number: MFCD00026126
CAS Number: 881-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR