6-(Cyanomethyl)nicotinonitrile ≥95%

Supplier: Matrix Scientific

056333-500MG 056333-1G
101910-076EA 346.15 USD
101910-076 101910-078
6-(Cyanomethyl)nicotinonitrile ≥95%
6-(Cyanomethyl)nicotinonitrile

Hazard: Irritant

Formula: C₈H₅N₃
MDL Number: MFCD09924285
CAS Number: 1000516-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR