5-Chloro-2-pyridinecarbohydrazide ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Chloropicolinohydrazide
056328-500MG
056328-1G
101910-056EA
384.62
USD
101910-056
101910-058
5-Chloro-2-pyridinecarbohydrazide ≥95%
5-Chloro-2-pyridinecarbohydrazide
Hazard: Irritant
Formula:
C₆H₆ClN₃O MW: 171.59 g/mol |
MDL Number:
MFCD16622817 CAS Number: 145835-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...