(2-Chloro-5-nitrophenyl)-N-methylmethanamine ≥95%
Supplier: Matrix Scientific
056326-500MG
056326-1G
056326-5G
101910-046EA
375.38
USD
101910-046
101910-048
101910-050
(2-Chloro-5-nitrophenyl)-N-methylmethanamine ≥95%
(2-Chloro-5-nitrophenyl)-N-methylmethanamine
Hazard: Irritant
Formula:
C₈H₉ClN₂O₂ |
MDL Number:
MFCD07686054 CAS Number: 872107-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...