4-Chloro-2,6-bis(trifluoromethyl)pyridine ≥95%
Supplier: Matrix Scientific
056319-500MG
056319-1G
101910-016EA
538.46
USD
101910-016
101910-018
4-Chloro-2,6-bis(trifluoromethyl)pyridine ≥95%
4-Chloro-2,6-bis(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₇H₂ClF₆N Melting Pt: 57…59 °C |
MDL Number:
MFCD00082558 CAS Number: 81269-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...