2'-Chloro[1,1'-biphenyl]-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

056318-500MG 056318-1G
101910-012EA 307.69 USD
101910-012 101910-014
2'-Chloro[1,1'-biphenyl]-2-carbaldehyde ≥95%
2'-Chloro[1,1'-biphenyl]-2-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₉ClO
MW: 216.67 g/mol
MDL Number: MFCD04039056
CAS Number: 223575-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR