5-Bromo-6-phenyl-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%

Supplier: Matrix Scientific

056315-500MG 056315-1G
101910-002EA 556.92 USD
101910-002 101910-004
5-Bromo-6-phenyl-1H-pyrazolo[3,4-b]pyridin-3-amine ≥95%
5-Bromo-6-phenyl-1H-pyrazolo[3,4-b]pyridin-3-amine

Hazard: Irritant

Formula: C₁₂H₉BrN₄
MDL Number: MFCD16622816
CAS Number: 583039-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR