5-Bromo-2-chloroisonicotinonitrile ≥95%

Supplier: Matrix Scientific

056313-500MG 056313-1G
101909-996EA 500 USD
101909-996 101909-998
5-Bromo-2-chloroisonicotinonitrile ≥95%
5-Bromo-2-chloroisonicotinonitrile

Hazard: Irritant

Formula: C₆H₂BrClN₂
MDL Number: MFCD16622809
CAS Number: 1211515-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR