4-Benzyl-1(2H)-phthalazinone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Benzylphthalazin-1(2H)-one

056311-500MG 056311-1G 056311-5G
101909-986EA 181.54 USD
101909-986 101909-988 101909-990
4-Benzyl-1(2H)-phthalazinone ≥95%
4-Benzyl-1(2H)-phthalazinone

Hazard: Irritant

Formula: C₁₅H₁₂N₂O
MDL Number: MFCD00139376
CAS Number: 32003-14-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR