1,3-Benzodioxole-5,6-diamine ≥95%

Supplier: Matrix Scientific

056310-500MG 056310-1G
101909-982EA 747.69 USD
101909-982 101909-984
1,3-Benzodioxole-5,6-diamine ≥95%
1,3-Benzodioxole-5,6-diamine

Hazard: Irritant/Store in Freezer

Formula: C₇H₈N₂O₂
MDL Number: MFCD07780137
CAS Number: 38608-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR