4-Amino-3-methoxybenzonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Cyano-2-methoxy-aniline
056309-500MG
056309-1G
101909-978EA
75.38
USD
101909-978
101909-980
4-Amino-3-methoxybenzonitrile ≥95%
4-Amino-3-methoxybenzonitrile
Hazard: Irritant
Formula:
C₈H₈N₂O MW: 148.1600 g/mol |
MDL Number:
MFCD13250177 CAS Number: 177476-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...