6-Bromo-3-chloropyridine-2-methanol ≥96%

Supplier: Matrix Scientific

056297-250MG 056297-1G
101909-960EA 112.31 USD
101909-960 101909-962
6-Bromo-3-chloropyridine-2-methanol ≥96%
6-Bromo-3-chloropyridine-2-methanol

Hazard: Irritant

Formula: C₆H₅BrClNO
MDL Number: MFCD16606992
CAS Number: 1227563-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR