6-Chloroimidazo[1,2-b]pyridazine ≥98%

Supplier: Matrix Scientific

056285-5G 056285-25G
101909-932EA 110.77 USD
101909-932 101909-934
6-Chloroimidazo[1,2-b]pyridazine ≥98%
6-Chloroimidazo[1,2-b]pyridazine

Hazard: Irritant

Formula: C₆H₄ClN₃
MW: 153.5691 g/mol
MDL Number: MFCD07778345
CAS Number: 6775-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR