3,6-Dichloropyridine-2-methanol ≥98%

Supplier: Matrix Scientific

Synonyms: 3,6-Dichloro-2-(hydroxymethyl)pyridine

056275-1G 056275-5G
101909-926EA 75.38 USD
101909-926 101909-928
3,6-Dichloropyridine-2-methanol ≥98%
3,6-Dichloropyridine-2-methanol

Hazard: Irritant

Formula: C₆H₅Cl₂NO
MW: 178.02 g/mol
MDL Number: MFCD13185536
CAS Number: 58804-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR