2-Amino-4-fluoropyridine ≥97%

Supplier: Matrix Scientific

055675-1G 055675-5G
101909-906EA 36.92 USD
101909-906 101909-908
2-Amino-4-fluoropyridine ≥97%
2-Amino-4-fluoropyridine

Hazard: Irritant

Formula: C₅H₅FN₂
MW: 112.04 g/mol
MDL Number: MFCD09260906
CAS Number: 944401-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR