5-Bromo-3-fluoropicolinonitrile

Supplier: Matrix Scientific

Synonyms: 5-bromo-3-fluoropyridine-2-carbonitrile

055564-1G 055564-5G
101909-684EA 2172.31 USD
101909-684 101909-686
5-Bromo-3-fluoropicolinonitrile
5-Bromo-3-fluoropicolinonitrile

Hazard: Irritant/Refrigerate

Formula: C₆H₂BrFN₂
MW: 200.9959 g/mol
MDL Number: MFCD07375030
CAS Number: 886373-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR