[4-(Trifluoromethyl)-2-pyridinyl]methanol ≥95%

Supplier: Matrix Scientific

Synonyms: (4-(Trifluoromethyl)pyridin-2-yl)methanol

055506-500MG 055506-1G
101909-562EA 484.62 USD
101909-562 101909-564
[4-(Trifluoromethyl)-2-pyridinyl]methanol ≥95%
[4-(Trifluoromethyl)-2-pyridinyl]methanol

Hazard: Irritant

Formula: C₇H₆F₃NO
MW: 177.13 g/mol
MDL Number: MFCD07774126
CAS Number: 131747-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR