[5-(Trifluoromethyl)-2-pyridinyl]methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Hydroxymethyl)-5-(trifluoromethyl)pyridine, (5-(Trifluoromethyl)pyridin-2-yl)methanol

055505-1G 055505-5G
101909-558EA 300 USD
101909-558 101909-560
[5-(Trifluoromethyl)-2-pyridinyl]methanol ≥95%
[5-(Trifluoromethyl)-2-pyridinyl]methanol

Hazard: Irritant

Formula: C₇H₆F₃NO
MW: 177.13 g/mol
MDL Number: MFCD04117762
CAS Number: 31181-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR