[3-Chloro-4-(trifluoromethyl)-2-pyridinyl]methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-2-(hydroxymethyl)-4-(trifluoromethyl)pyridine

055476-500MG 055476-1G
101909-452EA 1296.92 USD
101909-452 101909-454
[3-Chloro-4-(trifluoromethyl)-2-pyridinyl]methanol ≥95%
[3-Chloro-4-(trifluoromethyl)-2-pyridinyl]methanol

Hazard: Irritant

Formula: C₇H₅ClF₃NO
MW: 211.57 g/mol
MDL Number: MFCD16622807
CAS Number: 1228182-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR