2-(Trifluoromethyl)-3-azabicyclo[3.1.0]hexane ≥95%

Supplier: Matrix Scientific

055464-500MG 055464-1G
101909-396EA 1240 USD
101909-396 101909-398
2-(Trifluoromethyl)-3-azabicyclo[3.1.0]hexane ≥95%
2-(Trifluoromethyl)-3-azabicyclo[3.1.0]hexane

Hazard: Irritant

Formula: C₆H₈F₃N
MDL Number: MFCD14525504
CAS Number: 1212082-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR