(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: N-(tert-Butoxycarbonyl)-D-proline methyll ester, Boc-D-Pro-OMe, N-Boc-D-proline methyll ester, N-Boc-D-proline methyl ester

055463-500MG 055463-1G
101909-392EA 278.46 USD
101909-392 101909-394
(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate ≥95%
(R)-1-tert-Butyl-2-methyl pyrrolidine-1,2-dicarboxylate

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.27 g/mol
MDL Number: MFCD00190828
CAS Number: 73323-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR