3-Bromo-4-methylbenzotrifluoride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1-methyl-4-(trifluoromethyl)benzene

055424-1G 055424-5G
101909-232EA 23.08 USD
101909-232 101909-234
3-Bromo-4-methylbenzotrifluoride ≥97%
3-Bromo-4-methylbenzotrifluoride

Hazard: Irritant

Formula: C₈H₆BrF₃
MW: 239.04 g/mol
MDL Number: MFCD01631584
CAS Number: 66417-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR