3-Amino-1H-pyrazole-4-carboxamide sulfate(2:1) ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Aminopyrazole-4-carboxamide Hemisulphate, 3-Amino-4-pyrazolecarboxamide hemisulphate salt, 3-Amino-4-carbamoylpyrazole Hemisulphate, 3-Amino-1H-pyrazole-4-carboxamide hemisulphate, Allopurinol Related Compound A

055421-1G 055421-5G
101909-224EA 15.38 USD
101909-224 101909-226
3-Amino-1H-pyrazole-4-carboxamide sulfate(2:1) ≥95%
3-Amino-1H-pyrazole-4-carboxamide sulfate(2:1)

Hazard: Irritant

Formula: C8H14N8O6S
MDL Number: MFCD00013093
CAS Number: 27511-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR