2-Amino-3,5,6-trifluoropyridine ≥98%

Supplier: Matrix Scientific

055399-1G 055399-5G
101909-174EA 63.08 USD
101909-174 101909-176
2-Amino-3,5,6-trifluoropyridine ≥98%
2-Amino-3,5,6-trifluoropyridine

Hazard: Irritant

Formula: C₅H₃F₃N₂
MDL Number: MFCD03001164
CAS Number: 3534-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR