3,5-Dibromopicolinic acid ≥98%

Supplier: Matrix Scientific

054927-1G 054927-5G
101909-092EA 61.54 USD
101909-092 101909-094
3,5-Dibromopicolinic acid ≥98%
3,5-Dibromopicolinic acid

Hazard: Irritant

Formula: C₆H₃Br₂NO₂
MDL Number: MFCD06411110
CAS Number: 61830-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR