N-Isopropyl-2-(methylamino)acetamide hydrochloride

Supplier: Matrix Scientific

052850-500MG 052850-2.5G
101901-288EA 364.62 USD
101901-288 101901-290
N-Isopropyl-2-(methylamino)acetamide hydrochloride
N-Isopropyl-2-(methylamino)acetamide hydrochloride

Hazard: Irritant

Formula: C₆H₁₅ClN₂O
MDL Number: MFCD13196084
CAS Number: 1209744-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR