6-Chloro-N-isopropyl-2-pyridinamine

Supplier: Matrix Scientific

052848-500MG 052848-2.5G
101901-280EA 364.62 USD
101901-280 101901-282
6-Chloro-N-isopropyl-2-pyridinamine
6-Chloro-N-isopropyl-2-pyridinamine

Hazard: Irritant

Formula: C₈H₁₁ClN₂
MDL Number: MFCD00234439
CAS Number: 1220034-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR