5-Bromo-N-isopropyl-2-pyridinamine

Supplier: Matrix Scientific

052847-500MG 052847-2.5G
101901-276EA 364.62 USD
101901-276 101901-278
5-Bromo-N-isopropyl-2-pyridinamine
5-Bromo-N-isopropyl-2-pyridinamine

Hazard: Irritant

Formula: C₈H₁₁BrN₂
MDL Number: MFCD11122316
CAS Number: 443339-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR