5-Bromo-N-isopropyl-3-methyl-2-pyridinamine

Supplier: Matrix Scientific

052846-500MG 052846-2.5G
101901-272EA 364.62 USD
101901-272 101901-274
5-Bromo-N-isopropyl-3-methyl-2-pyridinamine
5-Bromo-N-isopropyl-3-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₉H₁₃BrN₂
MDL Number: MFCD13562406
CAS Number: 1219957-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR