N2-Isopropyl-2,3-pyridinediamine

Supplier: Matrix Scientific

Synonyms: N2-Isopropylpyridine-2,3-diamine

052844-500MG 052844-2.5G
101901-264EA 364.62 USD
101901-264 101901-266
N2-Isopropyl-2,3-pyridinediamine
N2-Isopropyl-2,3-pyridinediamine

Hazard: Irritant

Formula: C₈H₁₃N₃
MW: 151.21 g/mol
MDL Number: MFCD08692138
CAS Number: 24188-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR