2-(Isopropylamino)nicotinic acid

Supplier: Matrix Scientific

052843-500MG 052843-2.5G
101901-260EA 384.62 USD
101901-260 101901-262
2-(Isopropylamino)nicotinic acid
2-(Isopropylamino)nicotinic acid

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD04115113
CAS Number: 885275-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR